Horiba PH Laqua Pocket Tester, pH; Range 2 to 14 pH

Representative image only.
Horiba Mfr # 3200456563 – Item # EW-05754-10
Flat sensor enables easy measurement with only a few drops of sample!
  • User-friendly, three-button operation
  • Meters contain everything to get started
  • More than 200 hours continuous battery operation
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeTester
  • Display typeLCD
  • Min pH (pH)2
  • Max pH (pH)12
  • pH Resolution0.1
  • pH Accuracy±0.1 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 2 points
  • WaterproofYes
  • Battery1 x 3 V CR2025 battery (included)
  • Battery life (hr)200
  • Width (cm)2.8575
  • Height (cm)16.51
  • Width (in)1 1/8
  • Height (in)6 1/2
  • DescriptionLaqua Pocket Tester, pH; Range 2 to 14 pH
  • Warranty1 year
1 Year Warranty

Key Features

  • User-friendly, three-button operation
  • Meters contain everything to get started
  • More than 200 hours continuous battery operation
  • 1 year warranty

More About this Item

This pocket-sized IP67- rated waterproof sensor allows you to take simple, fast, and accurate measurements. Built in sensor lets you to take measurements in two ways: you can immerse the meter tip into sample, or you can place a few drops (0.3 mL) directly on the sensor. Results are instantly displayed on the 4-digit LCD. Automatic calibration function recognizes standards, and is user selectable for either one- or two-point calibrations.

Replacement sensor in accessories; 05754-51.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat