Horiba U-52G Multiparameter Meter, 33-ft (10-M) Cable

Representative image only.
Horiba Mfr # 3200164499 – Item # EW-99600-26
Measure and display pH, mV, conductivity, TDS, salinity, dissolved oxygen, turbidity, seawater specific gravity, water depth, and temperature simultaneously
  • Auto-calibration feature simplifies calibration of pH, dissolved oxygen, conductivity, turbidity, and depth
  • Built-in GPS — ideal for environmental surveys
  • Rugged housing is both waterproof and shock resistant
  • Stores up to 10,000 readings with USB connectivity for export of data to a PC
  • 1 year warranty

PRODUCT OPTIONS

$6,381.90USD / Each
$7,091.00USD / Each

$164.50 - $237.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.1 pH
  • Buffer RecognitionDIN, NIST, and USA
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution1 mV
  • mV Accuracy±15 mV
  • Min Conductivity0 mS
  • Max Conductivity100 mS
  • Conductivity Resolution0.001, 0.01, or 0.1 mS; 0.001 or 0.01 S
  • Conductivity Accuracy±1% FS
  • Min TDS0 to 100 ppt
  • Min Salinity0 to 70 ppt
  • Salinity Resolution0.1 ppt; 0.01%
  • Salinity Accuracy±3 ppt
  • Min Turbidity0 to 800 NTU
  • Turbidity Resolution0.1 NTU
  • Turbidity Accuracy±5% of reading or ±1 NTU
  • Min Dissolved Oxygen0 to 50 ppm
  • Dissolved Oxygen Resolution0.01 ppm
  • Dissolved Oxygen Accuracy±0.2 ppm
  • Min Temperature (° C)-5
  • Max Temperature (° C)50
  • Min Temperature (° F)23
  • Max Temperature (° F)122
  • Temperature compensationAutomatic
  • InterfaceUSB
  • Data Logging (points)10,000 data sets
  • Battery4 C (included)
  • Battery life (hr)70
  • Width (cm)11.43
  • Height (cm)28.2575
  • Width (in)4 1/2
  • Length (in)2 5/8
  • Height (in)11 1/8
  • DescriptionMultiparameter Meter, 33-ft (10-M) Cable
  • Warranty1 year
1 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier

Key Features

  • Auto-calibration feature simplifies calibration of pH, dissolved oxygen, conductivity, turbidity, and depth
  • Built-in GPS — ideal for environmental surveys
  • Rugged housing is both waterproof and shock resistant
  • Stores up to 10,000 readings with USB connectivity for export of data to a PC
  • 1 year warranty

More About this Item

These handheld, waterproof meters feature a large 3-1/2" (8.9 cm), easy-to-read LCD with backlighting for use in dark and low-light conditions. Multiple parameters can be measured and displayed at the same time. USB/probe connectivity, GPS info, and battery power are also indicated via the display. Variable contrast allows you to adjust for extreme ambient light conditions.

Meters are designed for one-handed operation. Probes are attached via a quick-connect connector allowing for fast setup. Unique auto-calibration feature provides hassle-free meter setup. Turbidity sensor offers better accuracy than most other conventional water quality meters. Polarographic DO sensor features improved stability, fast response, and easy maintenance.

Auto-hold function freezes average data values providing more time to verify or transcribe data. Up to 10,000 data sets can be stored and later transferred to a PC via a built-in USB connection. Units of measure are user selectable which helps you save time.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat