This item is now discontinued. We recommend the following as suggested replacements: EW-59200-40
Hydrion 7800 plastic pH indicator strips, 0.0 to 14.0, clear plastic case packaging
Representative image only.
Hydrion Mfr # 7800 – Item # EW-59200-10
Three times the matching area compared with other plastic strips
  • Easier to read and color match than other plastic strips
SPECS & DESCRIPTION
Specifications & Description
  • TestpH
  • Measurement Points1 to 14 pH
  • Reagent PlatformStrips
  • Tests Per Kit100
  • Descriptionplastic pH indicator strips, 0.0 to 14.0, clear plastic case packaging
Key Features
  • Easier to read and color match than other plastic strips
More About this Item
Immediate pH testing is available in brilliant, long lasting colors. These direct-reading pH papers give a simple, single color match for all pH values in distinct, bright color separations. Clear plastic cases contain 100 dip sticks and feature a handy matching color chart.