ICI Quick Ship Bench mounted Reagent Poles, 12" L

Representative image only.
ICI Quick Ship Mfr # CPRP-12 – Item # EW-78982-00
Customize shelving to fit your space
  • Create bench shelving
  • 1” diameter painted steel
  • Secure mounting
  • Stackable to create tiered shelving
  • Fastener on the reagent shelf surfaces is recessed
  • 2 year warranty

PRODUCT OPTIONS

$102.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Dimension1" diameter
  • Depth (in)63/64
  • Length (in)12
  • Depth (cm)2.5
  • MaterialSteel
  • ColorPearl white
  • DescriptionBench mounted Reagent Poles, 12" L
  • Warranty2 Years
Two year warrantyMade in the U.S.A.

Includes

Hardware for each installation

Key Features

  • Create bench shelving
  • 1” diameter painted steel
  • Secure mounting
  • Stackable to create tiered shelving
  • Fastener on the reagent shelf surfaces is recessed
  • 2 year warranty

More About this Item

Create an innovative storage space by customizing your shelving. ICI Quick Ship bench mounted reagent steel poles are a 1” diameter and are threaded at both ends. The poles bolt through the work surface for secure mounting. Poles are stackable to create tiered shelving in the center of your bench. All hardware is included for each installation. Order shelves separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat