ICI Quick Ship Black Phenolic Work Surface, 48" Wide, 30" D
Representative image only.
ICI Quick Ship Mfr # CPLP4830-1 – Item # EW-78984-62
Work surfaces for a quick delivery furniture program that builds complete labs
  • An extensive inventory of high quality furnishings, chemical fume hoods, and components
  • Clear and easy project communication
  • Design, project management, and factory trained installation
  • 2 year warranty
PRODUCT OPTIONS
$625.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBlack Phenolic Work Surface, 48\" Wide, 30\" D
  • Warranty2 Years
Two year warranty
Key Features
  • An extensive inventory of high quality furnishings, chemical fume hoods, and components
  • Clear and easy project communication
  • Design, project management, and factory trained installation
  • 2 year warranty
More About this Item
Custom manufactured work surfaces can be ordered in different depths, with special edge details, with extra high backsplashes, or a variety of cutouts. Choose from phenolic or epoxy material to meet your application needs.
Phenolic work surfaces have good chemical resistance to a wide range of acids, alkalies, and solvents. The 1” thick cellulose fiber reinforced phenolic resin core has a polyurethane copolymer surface. The appearance is black or grey with a smooth non-glare surface. This surface material is best whenever a high degree of chemical resistance is required.
Epoxy work surfaces also have a good chemical resistance to a wide range of acids, alkalies, and solvents. However, they also excel in resistance to heat and sturdiness. The 1” thick molded epoxy resin appears black with a smooth, non-porous surface. This surface material is best whenever resistance to heat is required.