ICI Quick Ship CHEM Exhaust Snorkel, 42.9" (109 cm) Radius
Representative image only.
ICI Quick Ship Mfr # CPCES-3 – Item # EW-78981-16
Safely eliminates heat and harmful fumes
  • Designed to be mounted from the wall or ceiling
  • Arm joints allow snorkel to rotate and swivel 360°
  • 2 year warranty
PRODUCT OPTIONS
$2,735.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Ductwork Diameter (in)3
  • DescriptionICI Quick Ship CHEM Exhaust Snorkel, 42.9\" (109 cm) Radius
  • Warranty2 Years
Two year warranty
Includes
Complete mounting components and hardware.
Key Features
  • Designed to be mounted from the wall or ceiling
  • Arm joints allow snorkel to rotate and swivel 360°
  • 2 year warranty
More About this Item
These lightweight, flexible exhaust snorkels with three adjustable, composite friction joints allow you to get close to the source of fumes and particles safely. The two outermost arm joints can rotate and swivel 360 degrees. Constructed of anodized aluminum, the snorkels offer strength, durability, and ease of handling. The arms are 3” in diameter and are available with a maximum working radius of 42.9” (109 cm) or 57.1” (145 cm). Each arm comes with a 5.5” mini hood that will accept optional hoods.
The CHEM model features a white electrophoretic deposition coating making it ideal for fume extraction in aggressive environments where corrosion resistance must be assured.
For sale in U.S. only!