ICI Quick Ship Laboratory Needle Valve, Four Turrets, Nitrogen
Representative image only.
ICI Quick Ship Mfr # CPL2880-134WSA-N – Item # EW-78985-09
Deliver flow control of laboratory gases to your ICI Quick Ship lab benches
  • Designed with deck-mounted turret base for easy flow control
  • Color-coded handles allow for easy gas identification
  • Choose from a wide range of configuration options suitable for any laboratory
  • Valve is constructed of forged brass with stainless steel floating need and replaceable seat
  • Designed with four turrets for nitrogen service
  • 2 year warranty
PRODUCT OPTIONS
$741.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsForged brass valve, floating stainless steel needle, replaceable stainless steel seat
  • Media CompatibilityNitrogen
  • Flow PatternFour 90° Turrets
  • DescriptionLaboratory Needle Valve, Four Turrets, Nitrogen
  • Warranty2 Years
Two year warranty
Key Features
  • Designed with deck-mounted turret base for easy flow control
  • Color-coded handles allow for easy gas identification
  • Choose from a wide range of configuration options suitable for any laboratory
  • Valve is constructed of forged brass with stainless steel floating need and replaceable seat
  • Designed with four turrets for nitrogen service
  • 2 year warranty
More About this Item
Laboratory service valves provide flow control of laboratory gases to your ICI Quick Ship lab setup. These brass service valves are available in both ball and needle valve options. Ball valves provide simple on/off control with a quarter turn of the knob. Needle valves allow for more gradual and precise control with proportional control of the knob. Both valve types are available in four flow pattern configurations—single, two at 90°, two at 180°, and four turrets at 90°. Choose from four service options including gas, air, vacuum, and nitrogen