ICI Quick Ship Wall mounted Cylinder Restraint, for Cylinder 4" to 14" dia
Representative image only.
ICI Quick Ship Mfr # CPCRWM – Item # EW-78986-22
Securely restrain gas cylinders
  • Holds one cylinder
  • 2 year warranty
PRODUCT OPTIONS
$329.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Tubes Held1
  • MaterialAluminum
  • DescriptionWall mounted Cylinder Restraint, for Cylinder 4\" to 14\" dia
  • Warranty2 Years
Two year warranty
Includes
Mounting hardware and nylon strap.
Key Features
  • Holds one cylinder
  • 2 year warranty
More About this Item
Safely secure your cylinders with either a wall mounted or free-standing cylinder restraint.
The wall mounted cylinder restraint is constructed of cast aluminum and includes mounting hardware and nylon strap. Several units can be placed adjacent to each other to secure a group of cylinders.
The free-standing cylinder restraints are constructed from 2” x 2” welded tubular steel and can hold anywhere from 2 to 9 cylinders. Security chains provide proper restraint of cylinders.
For sale in U.S. only!