Idec FL1D-B12RCE SmartRelay, 12/24VDC input, relay output, no display, 12/24 VDC

Representative image only.
Idec Mfr # FL1E-B12RCE – Item # EW-65524-02
Replace multiple timers and relays with one unit
  • Easy programming with on-board LCD
  • 48 maximum I/O points with expansion modules
  • FM approved for Class I, Division 2 Class 1, Zone 2 hazardous locations
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Load Rating12 to 24 VDC
  • Length (in)2 53/64
  • Height (in)3 35/64
  • Width (in)2 5/32
  • Length (cm)7.2
  • Height (cm)9
  • Width (cm)5.5
  • DescriptionSmartRelay, 12/24VDC input, relay output, no display, 12/24 VDC
  • Warranty1 Year
1 Year WarrantyCanadian Standards AssociationFM ApprovedUL Listed.

Key Features

  • Easy programming with on-board LCD
  • 48 maximum I/O points with expansion modules
  • FM approved for Class I, Division 2 Class 1, Zone 2 hazardous locations
  • 1 year warranty

More About this Item

Relays are an integral part of a process control system. They complete or interrupt a circuit or groups of circuits. These relays feature a real-time clock and calendar, plus 29 different function blocks for easy problem solving. Program and edit using the selection buttons or use the advanced exclusive software WindLGC. Use these programmable relays to manage automatic lighting, access control, watering systems, pump control, or ventilation systems. More advanced than a time delay relay, but less expensive than a PLC, the IDEC SmartRelay series fills the need for a compact, flexible control and monitoring device. Relays can replace multiple relays, timers, and counters in many applications.
The IDEC SmartRelay integrates programming and display functions into the control unit with an on-board display (LCD) and keypad. The LCD can display messages up to 40 characters long from a selection of 97 character types. The relays are also expandable,
with expansion modules that snap on with no special cables, tools, or programming. Select from three expansion modules with various I/O points to fit your need. A maximum of 56 function blocks and 8 internal relays can work at the same time. Order rewritable memory cartridge 65524-14 to save your program and duplicate it on other SmartRelays.
SmartRelays feature a compact body measuring only 2.83''L x 3.54''H x 2.16''W.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat