Idec RU2S-M-A24 General Purpose Relays; 2pdt ac24
Representative image only.
Idec Mfr # RU2S-M-A24 – Item # EW-65516-04
Two and four-pole lead-free, solder-free ice-cube relays provide control solutions
  • AC and DC coils
  • Snap -on marking plate
  • IP40 protection
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Load Rating250 VAC
  • Control Voltage24 VAC
  • Max Current (amps)10
  • DescriptionGeneral Purpose Relays; 2pdt ac24
  • Warranty1 Year
1 Year WarrantyCanadian Standards AssociationUL Listed.
Key Features
  • AC and DC coils
  • Snap -on marking plate
  • IP40 protection
  • 1 year warranty
More About this Item
Six and ten amp relays are available with two pole and four pole contacts and have color-coded manual latching levers. These low cost but well built relays have solder-free construction and no internal wires. Relays have a non-polarized green LED indicator for easy verification of operating status. Contact material is silver tin oxide indium for the two-pole relays and gold-silver alloy on silver for four-pole relays. Relay features contact schematic printed on body and snap-on marking plate.