Idex 6000-117 316 Stainless Steel Ferrule, 1/16" OD Tubing, 10-32 Coned; 50/PK
Representative image only.
Idex Mfr # 6000-117 – Item # EW-02022-24
Can withstand pressures up to 20,000 psi (1380 bar)!
  • Constructed of durable stainless steel
  • 10-32 coned configuration
  • For 1/16" OD tubing
  • 50/PK
SPECS & DESCRIPTION
Specifications & Description
  • Material316 stainless steel
  • Compatible WithNuts 02022-23 and -32
  • Description316 Stainless Steel Ferrule, 1/16\" OD Tubing, 10-32 Coned; 50/PK
Key Features
  • Constructed of durable stainless steel
  • 10-32 coned configuration
  • For 1/16" OD tubing
  • 50/PK
More About this Item
Use with IDEX high-pressure stainless steel nuts for your high-pressure applications. Stainless steel ferrules are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
Metal fittings should not be used with plastic ports because they can damage the ports.