Idex 8125-074 Stator Face Assembly for Valve Model 8125, Ceramic; 1/EA
Representative image only.
Idex Mfr # 8125-074 – Item # EW-02015-66
Prolong the life of your valves
  • Stators need replacement only if the ports or sealing surfaces become damaged
  • Use with valve model 8125
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCeramic
  • Compatible WithValve model 8125
  • DescriptionStator Face Assembly for Valve Model 8125, Ceramic; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Stators need replacement only if the ports or sealing surfaces become damaged
  • Use with valve model 8125
  • 1 year warranty
More About this Item
Optimize performance of your manual chromatography valves with these replacement stators.