Idex Stator for Valve Models 8125/8126, SS; 1/EA
Representative image only.
Idex Mfr # 8125-098 – Item # EW-02015-69
Prolong the life of your valves
  • Stators need replacement only if the ports or sealing surfaces become damaged
  • Use with valve models 8125/8126
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel
  • Compatible WithValve models 8125/8126
  • DescriptionStator for Valve Models 8125/8126, SS; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Stators need replacement only if the ports or sealing surfaces become damaged
  • Use with valve models 8125/8126
  • 1 year warranty
More About this Item
Optimize performance of your manual chromatography valves with these replacement stators.