Idex Rotor Seal for Valve Model 9010, ETFE, 0.015" Groove; 1/EA
Representative image only.
Idex Mfr # 9010-061 – Item # EW-02015-75
Replace rotor seals for longer valve life
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve model 9010
  • ETFE seal
  • 0.015" Groove
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialETFE
  • Compatible WithValve model 9010, 0.015\" Groove
  • DescriptionRotor Seal for Valve Model 9010, ETFE, 0.015\" Groove; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve model 9010
  • ETFE seal
  • 0.015" Groove
  • 1 year warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.