Idex 9085-20 ColumnSaver™ Direct-Connect Precolumn Filter, HPLC, PEEK, 2 µm SS Frit, 1/16" OD, 10-32 Coned; 1/EA
Representative image only.
Idex Mfr # 9085-20 – Item # EW-02010-19
Precolumn filters offer secure protection for analytical columns
  • Economical protection for larger columns and injections
  • Feature 10-32 coned ports for connection to 1/16" OD tubing
  • Replaceable stainless steel (SS) and PEEK® frits
  • Max pressure 6000 psi (414 bar)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPEEK
  • Housing MaterialStainless Steel
  • Pore Size (µm)2
  • Connections10-32 Coned
  • Max Pressure (PSI)6000
  • DescriptionColumnSaver™ Precolumn Filter, PEEK, 2 µm SS Frit, 1/16\" OD, 10-32 Coned; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Economical protection for larger columns and injections
  • Feature 10-32 coned ports for connection to 1/16" OD tubing
  • Replaceable stainless steel (SS) and PEEK® frits
  • Max pressure 6000 psi (414 bar)
  • 1 year warranty
More About this Item
Economical precolumn filters are designed to protect columns by filtering out particulate matter originating from the sample or from rotor seal wear. Filters feature 10-32 coned ports compatible with 1/16" outer diameter tubing.
This filter assembly is direct-connect for standard HPLC applications and requires polymer fittings.