Idex Iso-Prep Guard Column Cartridge, 2 µm Frit, 30 mm ID x 1 cm L; 1/EA

Representative image only.
Idex Mfr # 9196-20 – Item # EW-42712-10
Guard columns and hardware to meet your system requirements
  • Extend the life of analytical columns
  • Save on column replacement costs
  • Improves symmetry and plate count
  • 30 mm ID prep column protection
  • Includes 2 µm frit
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Pore Size (µm)2
  • Filter Diameter (mm)33
  • DescriptionIso-Prep Guard Column Cartridge, 2 µm Frit, 30 mm ID x 1 cm L
  • Warranty1 year
1 Year Warranty

Key Features

  • Extend the life of analytical columns
  • Save on column replacement costs
  • Improves symmetry and plate count
  • 30 mm ID prep column protection
  • Includes 2 µm frit
  • 1 year warranty

More About this Item

Protect the valuable columns in your system with guards, holders and analytical cartridges, among other hardware to help increase retention time, and promote excellent performance.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat