Idex MicroFilter Frit, SS, 10 µm, 0.750" OD, 0.880" OD ETFE Ring, 141.9 µL; 1/EA
Representative image only.
Idex Mfr # A-331 – Item # EW-12022-16
Extend filtration life and consistency with replaceable frits
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
PRODUCT OPTIONS
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialStainless Steel
  • Pore Size (µm)10
  • Filter Diameter (mm)0.88
  • DescriptionMicroFilter Frit, SS, 10 µm, 0.750\" OD, 0.880\" OD ETFE Ring, 141.9 µL; 1/EA
Key Features
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
More About this Item
Easily replaceable frits save you time and money lost to frit-cleaning labor, plus eliminate the problem of embedded debris re-entering your instrument. These frits are available in the most common HPLC filtration ratings. Each includes a PCTFE or PEEK polymer sealing ring, which are slightly thicker than the frit disc for extra sealing and chemical resistance. Many also have the most common outer diameters — 0.250" and 0.254" — making them ideal for use in inline or precolumn filters.