Idex A-346 Semi-Prep Frit, PEEK, 2 µm; 0.613" OD, 0.75" Green PCTFE Ring; 1/EA
Representative image only.
Idex Mfr # A-346 – Item # EW-02010-61
Extend filtration life and consistency with replaceable frits
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
  • Frit volume 85.5 µL
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPEEK
  • Pore Size (µm)2
  • Filter Diameter (mm)15.57
  • DescriptionSemi-Prep Frit, PEEK, 2 µm; 0.613\" OD, 0.75\" Green PCTFE Ring; 85.5 µL
  • Warranty1 year
1 Year Warranty
Key Features
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
  • Frit volume 85.5 µL
  • 1 year warranty
More About this Item
Easily replaceable frits save you time and money lost to frit-cleaning labor, plus eliminate the problem of embedded debris re-entering your instrument. These frits are available in the most common HPLC filtration ratings. Each includes a PCTFE or PEEK polymer sealing ring, which are slightly thicker than the frit disc for extra sealing and chemical resistance. Many also have the most common outer diameters — 0.250" and 0.254" — making them ideal for use in inline or precolumn filters.