Idex Adapter Fitting, Natural ETFE, 0.10" Bore, 3/16" Hose Barb x 1/4-28 Flat Bottom; 1/EA

Representative image only.
Idex Mfr # P-648 – Item # EW-02023-55
Easily connect soft-wall tubing to 10-32 coned, 1/4-28 flat-bottom, or 5/16-24 flat-bottom port
  • Choose from ETFE or PEEK construction
  • 3/16" hose barb x 1/4-28 flat bottom
  • 0.10" (2.5 mm) thru-hole diameter
  • 1 year warranty

PRODUCT OPTIONS

SPECS & DESCRIPTION

Specifications & Description

  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)1/4-28 Flat Bottom
  • Barb Size / Tubing Id (Port 2) (mm)4.8
  • Barb Size / Tubing Id (Port 2) (in)3/16
  • Compression Size / Tube Od (Port 1) (in)3/16
  • Compression Size / Tube Od (Port 1) (mm)4.763
  • ColorNatural
  • Orifice Dimensions (mm)2.5
  • DescriptionAdapter Fitting, Natural ETFE, 0.10" Bore, 3/16" Hose Barb x 1/4-28 Flat Bottom; 1/EA
  • Warranty1 year
1 Year Warranty

Key Features

  • Choose from ETFE or PEEK construction
  • 3/16" hose barb x 1/4-28 flat bottom
  • 0.10" (2.5 mm) thru-hole diameter
  • 1 year warranty

More About this Item

Slip soft-wall tubing over the barbed end then thread the adapter into a receiving port for a reliable low-pressure connection.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat