Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, PEEK, 2 µm, 3/16" OD Tubing; 1/EA
Representative image only.
Idex Mfr # A-436 – Item # EW-42711-41
Protect your system from damaging particulates
  • Filters are biocompatible—constructed of 100% PEEK polymer
  • 2 µm pore size
  • Use with 3/16" OD tubing
  • 1 year warranty
PRODUCT OPTIONS
$33.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPEEK
  • Pore Size (micron)2
  • Housing MaterialPEEK
  • Connections3/16" OD tubing
  • Typical Water Flow Rate30 mL/min (max)
  • DescriptionBottom-of-the-Bottle™ Biocompatible Filter Assembly, PEEK, 2 µm, 3/16" OD Tubing; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Filters are biocompatible—constructed of 100% PEEK polymer
  • 2 µm pore size
  • Use with 3/16" OD tubing
  • 1 year warranty
More About this Item
Filters allow solvents to be drawn within 0.080” (2.0mm) of the bottom of your solvent bottle. Use the 2 µm frit on the side for a 1/8” OD helium sparging line. Filters have 30 mL/min max flow rate.