Idex Flangeless Male Nut, Short Head, Red ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Mfr # P-318R – Item # EW-02020-08
Excellent for tight-space applications
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Red ETFE
  • 1/EA
  • 1 year warranty
PRODUCT OPTIONS
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorRed
  • DescriptionFlangeless Male Nut, Short Head, Red ETFE, 1/8\" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Red ETFE
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.