Idex Inlet Solvent Prep Filter, SS, 10 µm, 1/4-28 UNF; 1/EA
Representative image only.
Idex Mfr # A-311A – Item # EW-42711-22
Keep damaging particles out of your system with inlet solvent filters
  • Protect your system from particulate matter that can cause damage
  • Filters available to fit specific system specifications
  • Easy to replace without tools
  • Applications: High molecular weight hydrocarbons
  • 1 year warranty
PRODUCT OPTIONS
$98.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialStainless Steel
  • Pore Size (µm)10
  • Connections1/4-28 flat bottom
  • DescriptionInlet Solvent Filter, SS, 10 µm, 1/8\" OD Tubing; 1/EA
  • Warranty1 year
1 Year Warranty
Includes
One XP-315 ETFE flangeless fitting
Key Features
  • Protect your system from particulate matter that can cause damage
  • Filters available to fit specific system specifications
  • Easy to replace without tools
  • Applications: High molecular weight hydrocarbons
  • 1 year warranty
More About this Item
For good preventive maintenance, inlet solvent filters are an optimal way to cleanse solvents, avoiding pump cavitation and system damage. Different style stainless steel filters are available for specific system specifications and can be easily replaced regularly without tools.
To be used with 1/8" OD tubing.