Idex Inline Solvent Filter, UHMWPE, 10 µm, 1/16" OD, 1/4-28 Flat Bottom; 1/EA

Representative image only.
Idex Mfr # A-425 – Item # EW-02010-07
In-line Solvent Filters for running cleaner systems
  • Help prevent particulates from clogging equipment
  • Parts made of stainless steel and PEEK®
  • Replacement frits available
  • Applications: Alcohols, aromatics, flavors, free fatty acids, low-polarity pesticides
  • 1 year warranty

PRODUCT OPTIONS

$26.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Pore Size (µm)10
  • Connections1/4-28 Flat Bottom
  • Max Pressure (PSI)1000
  • DescriptionInline Solvent Filter, UHMWPE, 10 µm, 1/16" OD, 1/4-28 Flat Bottom; 1/EA
  • Warranty1 year
1 Year Warranty

Key Features

  • Help prevent particulates from clogging equipment
  • Parts made of stainless steel and PEEK®
  • Replacement frits available
  • Applications: Alcohols, aromatics, flavors, free fatty acids, low-polarity pesticides
  • 1 year warranty

More About this Item

In-line solvent filters are specifically engineered to help prevent particulate contamination from clogging sensitive equipment. Ideally, they should be placed along the flow path line between the pump and injection valve/autosampler. Available in a variety of porosities, filters threaded to fit 1/16" OD tubing, allowing the use of most standard chromatography high-pressure fittings.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat