Idex P-225 Flangeless Male Nut, Standard Knurl, Black ETFE, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
Representative image only.
Idex Mfr # P-225 – Item # EW-02019-64
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Black ETFE
  • 1/EA
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Thread Size ISO Metric (M) (Port 2) (mm)6
  • ColorBlack
  • DescriptionFlangeless Male Nut, Standard Knurl, Black ETFE, 1/16\" OD Tubing, M6 Flat-Bottom; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Removable and reusable—saving you money!
  • Black ETFE
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.