Idex Super Flangeless™ Nut, Standard Knurl, Natural PPS, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Mfr # P-281 – Item # EW-02021-71
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Standard knurl
  • Natural PPS
  • 1/EA
  • 1 year warranty
PRODUCT OPTIONS
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPPS
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/32
  • ColorNatural
  • DescriptionSuper Flangeless™ Nut, Standard Knurl, Natural PPS, 1/32\" or 1/16\" OD Tubing, 1/4-28 Flat Bottom; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Standard knurl
  • Natural PPS
  • 1/EA
  • 1 year warranty
More About this Item
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
Pressure rating of nut depends on ferrule used.