Idex P-642BLK Threaded Luer Adapter, Black ETFE, 0.050" Bore, Female Luer x Male 10-32 Coned; 1/EA
Representative image only.
Idex Mfr # P-642BLK – Item # EW-02023-54
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
  • Female Luer x Male 10-32 Coned
  • 0.050" thru-hole diameter
  • Manufactured from Black ETFE
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBlack ETFE
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Male 10-32 Coned
  • DescriptionThreaded Luer Adapter, Black ETFE, 0.050\" Bore, Female Luer x Male 10-32 Coned; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Manufactured from inert polymers
  • Female Luer x Male 10-32 Coned
  • 0.050" thru-hole diameter
  • Manufactured from Black ETFE
  • 1 year warranty
More About this Item
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.