Idex P-846-01 VacuTight™ Nut, Long, Headless, Red PEEK, 1/16" OD Tubing, 10-32 Flat Bottom; 1/EA
Representative image only.
Idex Mfr # P-846-01 – Item # EW-02017-49
Make dependable connections under vacuum and low pressure conditions
  • Small size ensures sufficient nut/thread engagement
  • Design can allow a large number of connections in a small space
  • Vacuum applications rated up to 25" Hg (84 kPa)
  • For 1/16" OD tubing
  • 1/4-28 flat-bottom thread
  • Headless, long body
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
1 Year Warranty
Key Features
  • Small size ensures sufficient nut/thread engagement
  • Design can allow a large number of connections in a small space
  • Vacuum applications rated up to 25" Hg (84 kPa)
  • For 1/16" OD tubing
  • 1/4-28 flat-bottom thread
  • Headless, long body
  • 1 year warranty
More About this Item
VacuTight fittings are ideal for high throughput screening and other automated liquid-handling applications due to their small size and dependable, airtight connections in low pressure and vacuum environments. The flat-bottom ferrule configuration impedes tubing inner diameter reduction and overcompression that can be a problem with many coned fittings.
All VacuTight nuts must be used only with VacuTight ferrules.