Idex P-872 Barbed Luer Adapter, Natural ETFE, 0.080" Bore, Female Luer Lock x 1/8" Hose Barb; 1/EA
Representative image only.
Idex Mfr # P-872 – Item # EW-02024-14
Excellent for connecting soft-wall tubing to a syringe or a low-pressure filter with luer connection
  • Choose from polypropylene or ETFE construction
  • Female Luer Lock x 1/8" Hose Barb
  • Manufactured from ETFE
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialETFE
  • Connection Type-Port 1Female luer lock
  • Barb Size-Port 2 (in)1/8
  • DescriptionLow-Pressure Adapter, Barbed, Natural ETFE, 0.080\" Bore, Female Luer Lock x 1/8\" Hose Barb; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose from polypropylene or ETFE construction
  • Female Luer Lock x 1/8" Hose Barb
  • Manufactured from ETFE
  • 1 year warranty
More About this Item
Female luer fittings come with a lock. Male luer fittings are available with slip or lock connection.