Idex PK-250X Super Flangeless™ Ferrule, VHP, PK/SS Ring, 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
Representative image only.
Idex Mfr # PK-250X – Item # EW-02022-11
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Max pressure of 10,000 psi (689 bar)
  • For 1/16" OD tubing
  • Manufactured from a proprietary PEEK blend (PK)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPK/SS
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • Max Operating Pressure (PSI)10000
  • DescriptionSuper Flangeless™ Ferrule, VHP, PK/SS Ring, 1/16\" OD Tubing, 1/4-28 Flat Bottom; 10/PK
  • Warranty1 year
1 Year Warranty
Key Features
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Max pressure of 10,000 psi (689 bar)
  • For 1/16" OD tubing
  • Manufactured from a proprietary PEEK blend (PK)
  • 1 year warranty
More About this Item
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.