Idex Rapid Replacement Pod for Actuated Valve 02016-07; 1/EA
Representative image only.
Idex Mfr # PD715-102 – Item # EW-02016-14
Regular maintenance ensures optimal valve performance
  • Optimize performance of your rotary shear valves
  • Use with valve 02016-07
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithActuated valve 02016-07
  • DescriptionRapid Replacement Pod for Actuated Valve 02016-07; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Optimize performance of your rotary shear valves
  • Use with valve 02016-07
  • 1 year warranty
More About this Item
Routine valve maintenance using RheBuild® kits or rapid replacement pods ensure optimal performance. The entire pod can be replaced quickly as a unit with virtually no instrument downtime for repair.