Idex Rotor Seal for Valve Models 7000L/7040L, Vespel®; 1/EA
Representative image only.
Idex Mfr # 7000-016 – Item # EW-02015-39
Replace rotor seals for longer valve life
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 7000L/7040L
  • Vespel® seal
  • 1 year warranty
PRODUCT OPTIONS
$157.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVespel®
  • Compatible WithValve models 7000L/7040L
  • DescriptionRotor Seal for Valve Models 7000L/7040L, Vespel®; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 7000L/7040L
  • Vespel® seal
  • 1 year warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.