Idex Rotor Seal for Valve Models 7125 and 7725, ETFE; 1/EA
Representative image only.
Idex Mfr # 7125-079 – Item # EW-02013-08
Replace rotor seals for longer valve life
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 7125 and 7725
  • ETFE seal
  • 1 month warranty
PRODUCT OPTIONS
$162.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialETFE
  • Compatible WithValve models 7125 and 7725
  • DescriptionRotor Seal for Valve Models 7125 and 7725, ETFE; 1/EA
1 Month Warranty
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 7125 and 7725
  • ETFE seal
  • 1 month warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.