Idex Sample Loop for 8125/8126 Valves, SS, 10 µL, 0.020" OD x 0.008" ID; 1/EA
Representative image only.
Idex Mfr # 8021 – Item # EW-02016-44
Square-cut, burr-free ends provide a firm, flush connection to valve ports
  • 316 stainless steel construction
  • High pressure resistance, low permeability
  • 10 µL sample volume
  • 1 year warranty
$111.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Wetted Materials316 stainless steel
  • Tubing Outside Diameter (OD) (in)0.02
  • Tubing Inside Diameter (ID) (mm)0.2
  • Total Volume Capacity (µL)10
  • DescriptionSample Loop for 8125/8126 Valves, SS, 10 µL, 0.020" OD x 0.008" ID; 1/EA
  • Warranty1 year
1 Year Warranty
Includes
Two fittings with ferrule.
Key Features
  • 316 stainless steel construction
  • High pressure resistance, low permeability
  • 10 µL sample volume
  • 1 year warranty
More About this Item
A sample loop is a defined length of tubing and fittings configured to match the angle of the injection valve ports. A sample is loaded and held in the loop until injection is triggered, either automatically or manually. Stainless steel loops are supplied with unswaged fittings, so the tube can be completely bottomed in the injection port before the ferrule is swaged. The loop volumes are nominal.