Idex 316 Stainless Steel Nut, 1/16" OD Tubing, M6 Coned; 10/PK
Representative image only.
Idex Mfr # U-450X – Item # EW-02022-40
Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar)!
  • Constructed of durable stainless steel
  • Applications: Cis and trans FAME isomers, dioxin analyses
  • 1 year warranty
PRODUCT OPTIONS
$150.25USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless steel
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)M6 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.588
  • Max Operating Pressure (PSI)20000
  • Description316 Stainless Steel Nut, 1/16\" OD Tubing, M6 Coned; 10/PK
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of durable stainless steel
  • Applications: Cis and trans FAME isomers, dioxin analyses
  • 1 year warranty
More About this Item
These 316 stainless steel nuts (and ferrules) are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
Metal fittings should not be used with plastic ports because they can damage the ports.