Idex UH-196X VHP UHPLC Fitting, Male Nut/Ferrule, SS, 1/16" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex Mfr # UH-196X – Item # EW-01940-22
Designed to withstand extreme pressures
  • Ideal for UHPLC applications Two compression points provide twice the grip of standard ferrule
  • 10-32 coned configuration
  • Requires 45 in-lb (5.08 N-m) of torque
  • 1/16" OD tubing
  • 10/PK
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.588
  • Compression Size / Tube Od (Port 2) (in)1/16
  • Max Operating Pressure (PSI)28800
  • DescriptionVHP UHPLC Fitting, Male Nut/Ferrule, SS, 1/16\" OD Tubing, 10-32 Coned; 10/PK
1 Month Warranty
Includes
Ferrules 02022-69.
Key Features
  • Ideal for UHPLC applications Two compression points provide twice the grip of standard ferrule
  • 10-32 coned configuration
  • Requires 45 in-lb (5.08 N-m) of torque
  • 1/16" OD tubing
  • 10/PK
  • 1 month warranty
More About this Item
The most demanding HPLC applications require fittings that will meet higher pressure requirements than standard chromatography fittings can accommodate. This patented line of VHP fittings are perfect for use in high-performance analytical systems. The unique double compression ferrule design does not restrict the inner diameter of tubing and allows a less concentrated bite on the tubing.