Idex UH-293X VHP Ferrule, SS, 1/32" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex Mfr # UH-293X – Item # EW-02007-70
Ferrules for use with VHP fittings
  • Designed to withstand extreme pressures
  • Ideal for UHPLC applications
  • Replacement ferrule for VHP SS fitting 02022-92
  • Max pressure 18,000 psi (1241 bar)
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSS
  • Compatible WithReplacement ferrule for VHP SS fitting 02022-92
  • DescriptionVHP Ferrule, SS, 1/32\" OD Tubing, 10-32 Coned; 10/PK
1 Month Warranty
Key Features
  • Designed to withstand extreme pressures
  • Ideal for UHPLC applications
  • Replacement ferrule for VHP SS fitting 02022-92
  • Max pressure 18,000 psi (1241 bar)
  • 1 month warranty
More About this Item
These fittings ensure reliable connections and performance for your instruments.