Idex VHP-556 Precolumn VHP Replacement Frit, SS, 0.2 µm; 1/EA
Representative image only.
Idex Mfr # VHP-556 – Item # EW-02010-41
Precolumn filters offer secure protection for analytical columns
  • Economical protection for larger columns and injections
  • Feature 10-32 coned ports for connection to 1/16" OD tubing
  • Replaceable stainless steel (SS) and PEEK® frits
  • Swept volume 1.8 µL
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialStainless Steel
  • Pore Size (µm)0.2
  • DescriptionPrecolumn VHP Replacement Frit, SS, 0.2 µm; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Economical protection for larger columns and injections
  • Feature 10-32 coned ports for connection to 1/16" OD tubing
  • Replaceable stainless steel (SS) and PEEK® frits
  • Swept volume 1.8 µL
  • 1 year warranty
More About this Item
Economical precolumn filters are designed to protect columns by filtering out particulate matter originating from the sample or from rotor seal wear. Filters feature 10-32 coned ports compatible with 1/16" outer diameter tubing.