Idex XP-220 Flangeless Fitting, Standard Knurl, Natural PP, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Mfr # XP-220 – Item # EW-02020-75
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Natural PP
  • 1/EA
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPP
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • Warranty1 year
1 Year Warranty
Includes
Blue ETFE ferrule 02019-33.
Key Features
  • Removable and reusable—saving you money!
  • Natural PP
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.