Idex XP-286X Flangeless Fitting, Headless, Natural PPS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Representative image only.
Idex Mfr # XP-286X – Item # EW-02014-98
Eliminate the need to flange tubing
  • "Headless" head is excellent for tight space applications
  • Natural PPS
  • For 1/16" OD tubing
  • 10/PK
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPPS
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
1 Month Warranty
Includes
Blue ETFE ferrules 02019-33.
Key Features
  • "Headless" head is excellent for tight space applications
  • Natural PPS
  • For 1/16" OD tubing
  • 10/PK
  • 1 month warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Removable and reusable components saves you time and money. Unlike the time required to flange tubing, it only takes a few seconds to change the components.