IKA 0020023167 Star Shaped Beater, Stainless Steel, 2000 mL
Representative image only.
IKA Mfr # 0020023167 – Item # EW-28711-59
Choose from a variety of vessels and other accessories to expand the capabilities of your IKA® grinding mill
  • Compatible with IKA grinder mill 28711-53
  • Suitable for fibrous materials, such as paper and plant material up to Mohs hardness 5
  • Compatible with IKA MultiDrive 250 (28711-55)
PRODUCT OPTIONS
$237.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Sample Capacity (mL)250
  • MaterialStainless steel
  • DescriptionStar Shaped Cutter, Stainless Steel
Two year warranty
Key Features
  • Compatible with IKA grinder mill 28711-53
  • Suitable for fibrous materials, such as paper and plant material up to Mohs hardness 5
  • Compatible with IKA MultiDrive 250 (28711-55)
More About this Item
Choose from a variety of cutters and beaters to expand the capabilities of your IKA MultiDrive grinding mill