IKA 2336000 Flexible Coupling for Glass Stirring Rods
Representative image only.
IKA Mfr # 0002336000 – Item # EW-50703-75
Get the most out of your IKA Mixer with these accessories
  • 2 year warranty
PRODUCT OPTIONS
$553.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFlexible coupling (Max torque: 14)
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
A variety of propellers, paddles, clamps, and stands are available.
Flexible coupling is required for strring tasks utilizing glass stirring rods, compensating for any structural variances. Clamping range of 1/4" to 2/5" (6-10 mm).