IKA 3703300 ULTRA TURRAX Ball Mill Tube BMT-20 G Tube, (25/pk)
Representative image only.
IKA Mfr # 0003703300 – Item # EW-04738-08
Choose the sample tube type and size to suit your application
  • Hygienic and clean homogenizing in closed tube system
  • High level of user safety
  • Process sample volumes from 2 - 50 mL
PRODUCT OPTIONS
$420.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)15
  • Capacity (Liters)0.015
  • DescriptionULTRA TURRAX Ball Mill Tube BMT-20 G Tube, (25/pk)
Key Features
  • Hygienic and clean homogenizing in closed tube system
  • High level of user safety
  • Process sample volumes from 2 - 50 mL
More About this Item
These unique disposable hermetically sealed tubes prevent sample contamination. Each sample tube contains a stirring device, blade, or ball for mixing and homogenizing small samples. Choose from three tube styles with different mixing devices to process your particular sample type.
Ball-Mill Tubes contain a glass ball or ceramic ball for dry milling of dry and brittle samples (e.g. kaolin, gypsum, colored pigments, tablets), cell maceration, and processing of materials mixed with fluids. Process sample volumes from 2 to 15 mL.