IKA 3710000 Platform with Rubber Mat Base for KS 4000i Incubated Shakers
Representative image only.
IKA Mfr # 3710000 – Item # EW-51715-54
Expand the uses of your shaker with these easy to use attachments.
  • For smooth shaking operations in the low viscosity range, e.g. for cell cultures, nutrient media in Petri dishes, culture bottles and vessels with a low center of gravity.
  • With integrated slip-resistant foil (PP).
PRODUCT OPTIONS
$333.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Use WithIKA KS4000 Incubating Shaker
  • Height (in)0.9
  • Width (in)18.5
  • Depth (in)17.5
  • DescriptionPlatform with Rubber Mat Base for KS 4000i Incubated Shakers
Key Features
  • For smooth shaking operations in the low viscosity range, e.g. for cell cultures, nutrient media in Petri dishes, culture bottles and vessels with a low center of gravity.
  • With integrated slip-resistant foil (PP).