IKA 4025100 (230V) Digital Block Heater for Use with 1 Aluminium Blocks 230V

Representative image only.
IKA Mfr # 0004025100 – Item # EW-36700-02
Precise temperature control for repeatable results
  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Number Of Blocks1
  • Max Temperature (° F)248
  • Max Temperature (° C)120
  • Power (VAC)230
  • Power (watts)150
  • Height (in)2 7/8
  • Width (in)8 63/64
  • Length (in)5 61/64
  • DescriptionDigital Block Heater for Use with 1 Aluminium Blocks 230V
  • Warranty2 year
Two year warranty

Key Features

  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes
  • 2 year warranty

More About this Item

Designed to ensure highly accurate and reproducible results, guaranteeing shorten heating times of samples. Dry heaters are ideal for determining melting and boiling points, enzyme reactions, incubation and activation of cultures, culture media tests, and in situ hybridizations.

IKA''s digital dry block heaters offer precise temperature control and rapid heat-up, and provide excellent stability to maintain results. The heaters feature a bright LCD and easy to use interface. Other features include the display of the actual temperature, user adjustable temperature set point, time display with set point, timer, counter, and external temperature probe.

Each heater accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes.Modular blocks are constructed from a solid aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. (order blocks separately below)

Single block dimensions (L x W x H):
3 3⁄4" x 3" x 2" (9.5 x 7.6 x 5.1 cm).
Double block dimensions (L x W x H):
6" x 3 3⁄4" x 2 1⁄4" (15.2 x 9.5 x 5.7 cm);
double block size fits 2/4/6-block heaters.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat