IKA LR 1000 Control Reaction Station; 115/230 VAC
Representative image only.
IKA Mfr # 0025001992 – Item # EW-28710-99
Optimize chemical reactions in your laboratory with these modular reaction systems
  • Digital display for monitoring speed and temperature
  • Included pH monitoring and weighing function (control model only)
  • RS-232 and USB connections for PC control (control model includes an option for Bluetooth)
  • Control package with wireless display
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Control TypeControl
  • Width (InchShort)17
  • Height (in)14
  • Max Temperature (° F)248
  • Max Temperature (° C)120
  • Power (VAC)115 / 230
  • Power (watts)1200
  • Combination Block -Tube / Vial Size AcceptedUp to 1000 mL
  • Depth (in)12
  • DescriptionControl System
  • Warranty3 Years
3 Year WarrantyIP54 Rated
Key Features
  • Digital display for monitoring speed and temperature
  • Included pH monitoring and weighing function (control model only)
  • RS-232 and USB connections for PC control (control model includes an option for Bluetooth)
  • Control package with wireless display
  • 3 year warranty
More About this Item
Both systems are used to reproduce and enhance chemical reactions as well as to mix, disperse, and homogenize. They have a wide range of uses from general laboratory to pharmaceutical. A digital display allows the user to reach a set temperature and speed in a controlled manner. Ports for output let you control and monitor your reaction right from your PC.
Control models present the addition of pH and weight measurement. Bluetooth interface and separate wireless controller added to increase convenience of use.