IKA TR 1 Tube Insert for Trayster, Holds 24 x 1.5 mL Tubes.
Representative image only.
IKA Mfr # 0004525100 – Item # EW-04304-34
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithIKA Trayster Overhead shaker
  • DescriptionTube Insert for Trayster, Holds 24 x 1.5 mL Tubes.
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Multiple accessory options expand the capacity and allow for different size tubes. Use multiple inserts to shake different tube sizes at one time.