Imada DB-1X Data Acquisition Software Data Button
Representative image only.
Imada Mfr # DB-1X – Item # EW-93955-20
Capture key data from your torque testers
  • Customized Excel spreadsheet designed for capturing and analyzing peak and first peak data
  • Ideal for torque testers with either RS-232 or USB ports
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionData Acquisition Software Data Button
  • Warranty2 year
Two year warranty
Key Features
  • Customized Excel spreadsheet designed for capturing and analyzing peak and first peak data
  • Ideal for torque testers with either RS-232 or USB ports
  • 2 year warranty
More About this Item
Gather peak torque or first peak readings as well as a running log of all torque measurements along with a chart of the data and statistical information. All data is entered directly into a standard Excel spreadsheet, allowing you to analyze your torque tester data using all of the powerful tools available in Excel.
DATA BUTTON FOR DS2, DTX2, HTG2, ZP, ZPH
Use with SW-1 data acquisition software to capture and display the current force values. Press the button to capture values and zero the gauge.
DS2 REQUIRES CB-203 CABLE
DTX2, HTG2, ZP, ZPH REQUIRE CB-204 CABLE