Imada EX-C Asker C:Durometer Ex W/Hemi Type Tip For Soft Rubber And Sponge
Representative image only.
Imada Mfr # EX-C – Item # EW-59790-05
Won’t destroy your sample
  • Heavy-duty units feature well-made design with high repeatability
  • Measurement scale is 100 points with an accuracy of ±1 point
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • TypeAsker C
  • Warranty2 year
Two year warranty
Key Features
  • Heavy-duty units feature well-made design with high repeatability
  • Measurement scale is 100 points with an accuracy of ±1 point
  • 2 year warranty
More About this Item
Durometers feature portable design, ease of operation, and non-destructive measurement. They are heavy-duty and well-made with high repeatability. The measurement of hardness is an essential factor for determining the characteristics of various rubber, elastomers and plastic products. One of the most important steps in using a hardness tester is to select the optimum type from the various types available based upon different material hardnesses. The measurement scale is 100 points with an accuracy of ± 1 point.
Hardness Tester Selection
  • Model D: Hard rubber and plastics
  • Model A: Normal rubber and softer plastics
  • Asker C: Soft rubber and sponge