Imada HTGS-85 Torque Gauge 85LBF Capacity Digital
Representative image only.
Imada Mfr # HTGS-85 – Item # EW-59850-30
  • 6.5 mm Jacobs Chuck
PRODUCT OPTIONS
$1,476.56USD / Each
$1,707.00USD / Each
$250.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lb-in)85
  • Capacity (kg-mm)10
  • Capacity (N-cm)100
  • Resolution (lb-in)0.1
  • Resolution (kg-mm)0.1
  • Resolution (N-cm)1
  • Accuracy±0.5% F.S. ±1 LSD
  • OutputRS-232, Digimatic, and analog
  • Overload200% overload capacity
  • DisplayLCD
  • Capacity85 lb in
  • DescriptionTorque Gauge 85LBF Capacity Digital
Key Features
  • 6.5 mm Jacobs Chuck
More About this Item
FEATURES
  • Programmable high/low setpoints with LED indicators for quick GO/NO GO testing
  • 256 Data memory for recall and download
  • Accuracy: ±0.5% F.S. ±1 LSD
  • lbf-in, kgf-cm, and N-cm (selectable)
  • Outputs: RS-232, Digimatic, analog
  • 200% overload capacity
  • Ergonomic, heavy duty metal case
  • Real time and peak mode (selectable)
  • Tare Function
  • Clockwise and counterclockwise operation
  • Runs on internal NiCad batteries or AC adapter
  • Overload alarm, low battery indicator
  • Carrying case, standard attachments and AC adapter/charger included