Imada SW-2X Data Acquisition Software for Force and Displacement
Representative image only.
Imada Mfr # SW-2X – Item # EW-59850-49
Capture key data from your torque testers
  • Customized Excel spreadsheet designed for capturing and analyzing peak and first peak data
  • Ideal for torque testers with either RS-232 or USB ports
  • Full SPC and graphing capabilities
  • Calculates statistical data
  • Requires DB-2 data button and distance meter cable (905691)
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionData Acquisition Software for Force and Displacement
Two year warranty
Key Features
  • Customized Excel spreadsheet designed for capturing and analyzing peak and first peak data
  • Ideal for torque testers with either RS-232 or USB ports
  • Full SPC and graphing capabilities
  • Calculates statistical data
  • Requires DB-2 data button and distance meter cable (905691)
  • 2 year warranty
More About this Item
Gather peak torque or first peak readings as well as a running log of all torque measurements along with a chart of the data and statistical information. All data is entered directly into a standard Excel spreadsheet, allowing you to analyze your torque tester data using all of the powerful tools available in Excel.
Use SW-2 data acquisition software to capture and display the current force and displacement values and calculate max/min, average, and standard deviation. Use buttons on the spreadsheet to operate the gauge from your computer. Compatible with test stands equipped with distance meter.